Assay Details
| Gene Symbol: CAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161282.1 | NP_001154754.1 | 3 | 762 | 99 | |
| NM_001161283.1 | NP_001154755.1 | 3 | 795 | 99 | ||
| NM_001344219.1 | NP_001330399.1 | 2 | 874 | 99 | ||
| NM_123137.4 | NP_198594.1 | 2 | 779 | 99 | ||
More Information
Set Membership: |
|

CAM1
Genomic Map