Assay Details
| Gene Symbol: MAP18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344568.1 | NP_001331522.1 | 2 | 861 | 83 | |
| NM_001344569.1 | NP_001331521.1 | 2 | 527 | 83 | ||
| NM_123828.4 | NP_568636.1 | 2 | 621 | 83 | ||
| GenBank mRNA | AK117253.1 | - | 2 | 567 | 83 | |
More Information
Set Membership: |
|

MAP18
Genomic Map