Assay Details
| Gene Symbol: ZFP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102305.2 | NP_564223.1 | 2 | 850 | 85 | |
| GenBank mRNA | AK119018.1 | - | 2 | 831 | 85 | |
| BT005629.1 | - | 1 | 575 | 85 | ||
| L39650.1 | - | 2 | 851 | 85 | ||
More Information
Set Membership: |
|

ZFP7
Genomic Map