Assay Details
| Gene Symbol: SOFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102385.2 | NP_564241.1 | 2 | 1024 | 98 | |
| GenBank mRNA | AK175479.1 | - | 2 | 823 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOFL1| Gene Symbol: SOFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102385.2 | NP_564241.1 | 2 | 1024 | 98 | |
| GenBank mRNA | AK175479.1 | - | 2 | 823 | 98 | |
Set Membership: |
|