Assay Details
| Gene Symbol: NMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125084.3 | NP_568846.1 | 2 | 1646 | 162 | |
| GenBank mRNA | AF193616.1 | - | 2 | 1646 | 162 | |
| AF250956.1 | - | 2 | 1523 | 162 | ||
| AY045854.1 | - | 2 | 1551 | 162 | ||
More Information
Set Membership: |
|

NMT1
Genomic Map