Assay Details
| Gene Symbol: SIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125224.3 | NP_568878.1 | 2 | 1953 | 131 | |
| GenBank mRNA | AK226938.1 | - | 2 | 1746 | 131 | |
| AY094414.1 | - | 1 | 1476 | 131 | ||
| AY124002.1 | - | 2 | 1844 | 131 | ||
More Information
Set Membership: |
|

SIP1
Genomic Map