Assay Details
| Gene Symbol: BAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125967.5 | NP_201371.1 | 2 | 3273 | 121 | |
| GenBank mRNA | AK226485.1 | - | 2 | 3158 | 121 | |
| AY099814.1 | - | 2 | 3141 | 121 | ||
More Information
Set Membership: |
|

BAM1
Genomic Map