Assay Details
| Gene Symbol: AT2G28690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128430.3 | NP_180437.2 | 2 | 741 | 65 | |
| GenBank mRNA | AY102556.1 | - | 2 | 726 | 65 | |
| AY649292.1 | - | 2 | 575 | 65 | ||
More Information
Set Membership: |
|

AT2G28690
Genomic Map