Assay Details
| Gene Symbol: TIM17-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129296.4 | NP_181277.1 | 2 | 1144 | 105 | |
| GenBank mRNA | AF360258.1 | - | 2 | 1044 | 105 | |
| AY056107.1 | - | 2 | 1019 | 105 | ||
More Information
Set Membership: |
|

TIM17-2
Genomic Map