Assay Details
| Gene Symbol: AT2G39170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129474.3 | NP_181450.1 | 2 | 924 | 89 | |
| GenBank mRNA | BT003866.1 | - | 2 | 837 | 89 | |
| BT005082.1 | - | 1 | 674 | 89 | ||
More Information
Set Membership: |
|

AT2G39170
Genomic Map