Assay Details
| Gene Symbol: OXS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129754.4 | NP_565962.1 | 2 | 2575 | 102 | |
| GenBank mRNA | AY037232.1 | - | 2 | 2433 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OXS2| Gene Symbol: OXS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129754.4 | NP_565962.1 | 2 | 2575 | 102 | |
| GenBank mRNA | AY037232.1 | - | 2 | 2433 | 102 | |
Set Membership: |
|