Assay Details
| Gene Symbol: AT2G43535 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129915.4 | NP_566000.1 | 2 | 498 | 93 | |
| GenBank mRNA | AK228120.1 | - | 2 | 396 | 93 | |
| Gene Symbol: SS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK230236.1 | - | 1207 | 93 | ||
More Information
Set Membership: |
|

AT2G43535
Genomic Map