Assay Details
| Gene Symbol: SUVH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179699.3 | NP_850030.1 | 2 | 2248 | 65 | |
| NM_201785.1 | NP_973514.1 | 2 | 2191 | 65 | ||
| GenBank mRNA | BT002751.1 | - | 1 | 2066 | 65 | |
More Information
Set Membership: |
|

SUVH6
Genomic Map