Assay Details
| Gene Symbol: CAM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198401.1 | NP_001185330.1 | 3 | 708 | 165 | |
| NM_001334237.1 | NP_001320584.1 | 2 | 1449 | 165 | ||
| NM_001334238.1 | NP_001320585.1 | 2 | 824 | 165 | ||
| NM_105312.4 | NP_176814.1 | 2 | 638 | 165 | ||
| Gene Symbol: CAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF370293.1 | - | 2 | 530 | 165 | |
| AY088476.1 | - | 2 | 536 | 165 | ||
More Information
Set Membership: |
|

CAM4
Genomic Map