Assay Details
| Gene Symbol: SUVH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105968.3 | NP_565056.1 | 3 | 2289 | 106 | |
| GenBank mRNA | AY099620.1 | - | 3 | 2249 | 106 | |
| BT002137.1 | - | 1 | 2078 | 106 | ||
More Information
Set Membership: |
|

SUVH3
Genomic Map