Assay Details
| Gene Symbol: SAMDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084624.1 | NP_001078093.1 | 4 | 1664 | 129 | |
| NM_001161113.1 | NP_001154585.1 | 5 | 1726 | 129 | ||
| NM_111114.2 | NP_186896.1 | 3 | 1746 | 129 | ||
| GenBank mRNA | AF428468.1 | - | 4 | 1673 | 129 | |
| AK316891.1 | - | 4 | 1677 | 129 | ||
| AY042824.1 | - | 4 | 1673 | 129 | ||
| AY081446.1 | - | 1 | 1132 | 129 | ||
More Information
Set Membership: |
|

SAMDC
Genomic Map