Assay Details
| Gene Symbol: GSTF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126346.3 | NP_178394.1 | 2 | 569 | 143 | |
| GenBank mRNA | AF288181.1 | - | 2 | 354 | 143 | |
| BT024921.1 | - | 2 | 354 | 143 | ||
More Information
Set Membership: |
|

GSTF3
Genomic Map