Assay Details
| Gene Symbol: MYB114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334235.1 | NP_001321376.1 | 2 | 477 | 108 | |
| NM_001334236.1 | NP_001321375.1 | 2 | 290 | 108 | ||
| NM_105309.4 | NP_176812.1 | 3 | 451 | 108 | ||
| GenBank mRNA | AY008379.1 | - | 3 | 336 | 108 | |
| AY519567.1 | - | 3 | 336 | 108 | ||
| DQ446401.1 | - | 3 | 336 | 108 | ||
More Information
Set Membership: |
|

MYB114
Genomic Map