Assay Details
| Gene Symbol: TPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100132.2 | NP_171752.1 | 1 | 683 | 126 | |
| GenBank mRNA | AY258073.1 | - | 1 | 684 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPK4| Gene Symbol: TPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100132.2 | NP_171752.1 | 1 | 683 | 126 | |
| GenBank mRNA | AY258073.1 | - | 1 | 684 | 126 | |
Set Membership: |
|