Assay Details
| Gene Symbol: DRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101253.3 | NP_172840.1 | 2 | 1080 | 132 | |
| GenBank mRNA | BT004091.1 | - | 2 | 1056 | 132 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DRL1| Gene Symbol: DRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101253.3 | NP_172840.1 | 2 | 1080 | 132 | |
| GenBank mRNA | BT004091.1 | - | 2 | 1056 | 132 | |
Set Membership: |
|