Assay Details
| Gene Symbol: TOM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103831.5 | NP_564545.1 | 1 | 177 | 79 | |
| GenBank mRNA | AF324714.2 | - | 1 | 69 | 79 | |
| AF325059.2 | - | 1 | 115 | 79 | ||
| AF327425.1 | - | 1 | 69 | 79 | ||
| AF339720.1 | - | 1 | 45 | 79 | ||
| AY037189.1 | - | 1 | 69 | 79 | ||
| BT002672.1 | - | 1 | 45 | 79 | ||
More Information
Set Membership: |
|

TOM6
Genomic Map