Assay Details
| Gene Symbol: AT4G14320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117509.3 | NP_193168.1 | 4 | 524 | 91 | |
| Gene Symbol: AT3G23390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY085784.1 | - | 4 | 485 | 91 | |
More Information
Set Membership: |
|

AT4G14320
Genomic Map