Assay Details
| Gene Symbol: VHA-C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342512.1 | NP_001329869.1 | 3 | 1266 | 81 | |
| NM_120052.3 | NP_195603.1 | 3 | 927 | 81 | ||
| Gene Symbol: AT2G16510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF412097.1 | - | 3 | 734 | 81 | |
| AY049249.1 | - | 3 | 735 | 81 | ||
| AY059836.1 | - | 3 | 743 | 81 | ||
| AY087112.1 | - | 3 | 734 | 81 | ||
More Information
Set Membership: |
|

VHA-C3
Genomic Map