Assay Details
| Gene Symbol: RANGAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343596.1 | NP_001318597.1 | 3 | 1991 | 135 | |
| NM_121937.5 | NP_197433.1 | 2 | 1880 | 135 | ||
More Information
Set Membership: |
|

RANGAP2
Genomic Map