Assay Details
| Gene Symbol: SUVR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202656.2 | NP_001189585.1 | 6 | 3738 | 99 | |
| NM_001335861.1 | NP_001324207.1 | 6 | 3619 | 99 | ||
| NM_001335862.1 | NP_001324206.1 | 6 | 3479 | 99 | ||
| NM_001335863.1 | NP_001324205.1 | 6 | 3707 | 99 | ||
| NM_001335864.1 | NP_001324204.1 | 6 | 3520 | 99 | ||
| NM_127937.2 | NP_179954.2 | 6 | 3637 | 99 | ||
| GenBank mRNA | AK318822.1 | - | 6 | 3658 | 99 | |
More Information
Set Membership: |
|

SUVR5
Genomic Map