Assay Details
| Gene Symbol: SDR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130281.5 | NP_182235.1 | 2 | 669 | 62 | |
| GenBank mRNA | AK175374.1 | - | 2 | 558 | 62 | |
| BT010410.1 | - | 1 | 500 | 62 | ||
More Information
Set Membership: |
|

SDR3
Genomic Map