Assay Details
| Gene Symbol: CAM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180529.4 | NP_850860.1 | 2 | 600 | 113 | |
| GenBank mRNA | AY050350.1 | - | 2 | 566 | 113 | |
| Z12024.1 | - | 533 | 111 | |||
More Information
Set Membership: |
|

CAM6
Genomic Map