Assay Details
| Gene Symbol: AT1G03982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035876.2 | NP_001030953.1 | 2 - 3 | 795 | 91 | |
| GenBank mRNA | DQ487432.1 | - | 2 - 3 | 212 | 91 | |
More Information
Set Membership: |
|

AT1G03982
Genomic Map