Assay Details
Gene Symbol: AT1G58055 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036127.3 | NP_001031204.1 | 1 - 2 | 109 | 115 | |
GenBank mRNA | DQ912209.1 | - | 1 - 2 | 84 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
