Assay Details
| Gene Symbol: AT1G63522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334104.1 | NP_001319305.1 | 2 | 201 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G63522| Gene Symbol: AT1G63522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334104.1 | NP_001319305.1 | 2 | 201 | 100 | |
Set Membership: |
|