Assay Details
Gene Symbol: AT2G04046 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036240.2 | NP_001031317.1 | 1 - 2 | 51 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AT2G04046 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036240.2 | NP_001031317.1 | 1 - 2 | 51 | 73 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |