Assay Details
| Gene Symbol: CDKB1;2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036430.3 | NP_001031507.1 | 1 - 2 | 709 | 101 | |
| NM_001336710.1 | NP_001324504.1 | 1 - 2 | 709 | 101 | ||
| NM_001336711.1 | NP_001324503.1 | 2 - 3 | 479 | 101 | ||
| NM_129419.3 | NP_181396.2 | 1 - 2 | 677 | 101 | ||
| GenBank mRNA | AJ297937.1 | - | 1 - 2 | 631 | 101 | |
More Information
Set Membership: |
|

CDKB1;2
Genomic Map