Assay Details
| Gene Symbol: RIC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036663.2 | NP_001031740.1 | 2 - 3 | 833 | 132 | |
| NM_001341923.1 | NP_001329671.1 | 1 - 2 | 133 | 132 | ||
| GenBank mRNA | DQ487576.1 | - | 2 - 3 | 183 | 132 | |
More Information
Set Membership: |
|

RIC7
Genomic Map