Assay Details
Gene Symbol: SCRL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036706.2 | NP_001031783.1 | 1 - 2 | 120 | 116 | |
NM_001342216.1 | NP_001328489.1 | 1 - 2 | 120 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
