Assay Details
Gene Symbol: AT5G16453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036814.3 | NP_001031891.1 | 1 - 2 | 144 | 89 | |
GenBank mRNA | DQ912223.1 | - | 1 - 2 | 64 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
