Assay Details
| Gene Symbol: AT5G19485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036831.4 | NP_001031908.1 | 10 - 11 | 1266 | 62 | |
| GenBank mRNA | DQ487627.1 | - | 10 - 11 | 1084 | 62 | |
More Information
Set Membership: |
|

AT5G19485
Genomic Map