Assay Details
Gene Symbol: AT5G35195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036890.1 | NP_001031967.1 | 1 | 135 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AT5G35195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036890.1 | NP_001031967.1 | 1 | 135 | 105 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |