Assay Details
Gene Symbol: AT5G43401 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036924.1 | NP_001032001.1 | 1 - 2 | 65 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AT5G43401 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036924.1 | NP_001032001.1 | 1 - 2 | 65 | 76 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |