Assay Details
| Gene Symbol: NTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340330.1 | NP_001329585.1 | 4 - 5 | 1224 | 105 | |
| NM_116384.3 | NP_192063.2 | 4 - 5 | 1224 | 105 | ||
| GenBank mRNA | AB493671.1 | - | 4 - 5 | 1064 | 105 | |
More Information
Set Membership: |
|

NTM1
Genomic Map