Assay Details
| Gene Symbol: NTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340331.1 | NP_001329584.1 | 4 - 5 | 1453 | 123 | |
| NM_116384.3 | NP_192063.2 | 5 - 6 | 1371 | 123 | ||
| GenBank mRNA | AB493671.1 | - | 5 - 6 | 1211 | 123 | |
More Information
Set Membership: |
|

NTM1
Genomic Map