Assay Details
| Gene Symbol: FRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203847.2 | NP_001190776.1 | 4 - 5 | 2386 | 126 | |
| NM_001341387.1 | NP_001320002.1 | 4 - 5 | 2447 | 126 | ||
| NM_001341388.1 | NP_001328358.1 | 4 - 5 | 2421 | 126 | ||
| NM_001341389.1 | NP_001328360.1 | 3 - 4 | 2643 | 126 | ||
| NM_001341390.1 | NP_001328359.1 | 3 - 4 | 2643 | 126 | ||
| NM_001341391.1 | NP_001328362.1 | 3 - 4 | 2643 | 126 | ||
| NM_001341392.1 | NP_001328361.1 | 4 - 5 | 2534 | 126 | ||
More Information
Set Membership: |
|

FRS1
Genomic Map