Assay Details
| Gene Symbol: AT5G50530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124440.3 | NP_568736.1 | 1 - 2 | 209 | 135 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_124440.3 transcripts.
Set Membership: |
|

AT5G50530
Genomic Map