Assay Details
| Gene Symbol: AGL95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127127.3 | NP_179168.2 | 1 | 1120 | 147 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL95| Gene Symbol: AGL95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127127.3 | NP_179168.2 | 1 | 1120 | 147 | |
Set Membership: |
|