Assay Details
| Gene Symbol: NF-YC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_202067.2 | NP_973796.1 | 2 - 3 | 936 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NF-YC9| Gene Symbol: NF-YC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_202067.2 | NP_973796.1 | 2 - 3 | 936 | 101 | |
Set Membership: |
|