Assay Details
| Gene Symbol: ACP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104340.5 | NP_564663.1 | 3 - 4 | 530 | 108 | |
| GenBank mRNA | AY054604.1 | - | 3 - 4 | 356 | 108 | |
| AY072527.1 | - | 3 - 4 | 283 | 108 | ||
More Information
Set Membership: |
|

ACP3
Genomic Map