Assay Details
| Gene Symbol: UBC35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036221.3 | NP_001031298.1 | 5 - 6 | 401 | 91 | |
| NM_001334850.1 | NP_001319409.1 | 4 - 5 | 593 | 91 | ||
| NM_001334851.1 | NP_001321618.1 | 4 - 5 | 630 | 91 | ||
| NM_106535.4 | NP_565192.1 | 5 - 6 | 459 | 91 | ||
| GenBank mRNA | AY049261.1 | - | 5 - 6 | 356 | 91 | |
| AY085854.1 | - | 5 - 6 | 379 | 91 | ||
| BT000544.1 | - | 5 - 6 | 246 | 91 | ||
| DQ027048.1 | - | 5 - 6 | 246 | 91 | ||
More Information
Set Membership: |
|

UBC35
Genomic Map