Assay Details
| Gene Symbol: FUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340534.1 | NP_001329797.1 | 1 - 2 | 129 | 114 | |
| NM_116751.4 | NP_192421.1 | 2 - 3 | 260 | 114 | ||
| GenBank mRNA | AF426400.1 | - | 1 - 2 | 36 | 114 | |
| AK175753.1 | - | 2 - 3 | 117 | 114 | ||
More Information
Set Membership: |
|

FUR1
Genomic Map