Assay Details
| Gene Symbol: RPS5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036425.2 | NP_001031502.1 | 1 - 2 | 272 | 96 | |
| NM_129283.4 | NP_181264.1 | 2 - 3 | 446 | 96 | ||
| GenBank mRNA | AY059849.1 | - | 2 - 3 | 193 | 96 | |
| AY081669.1 | - | 1 - 2 | 96 | 96 | ||
More Information
Set Membership: |
|

RPS5B
Genomic Map