Assay Details
| Gene Symbol: AGL90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122679.2 | NP_198148.2 | 1 - 2 | 656 | 147 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL90| Gene Symbol: AGL90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122679.2 | NP_198148.2 | 1 - 2 | 656 | 147 | |
Set Membership: |
|