Assay Details
| Gene Symbol: AT5G53750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124754.4 | NP_200186.2 | 1 - 2 | 622 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G53750| Gene Symbol: AT5G53750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124754.4 | NP_200186.2 | 1 - 2 | 622 | 117 | |
Set Membership: |
|